Assay Details
| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024646.2 | NP_078922.1 | 1 - 2 | 228 | 123 | |
| XM_017002336.1 | XP_016857825.1 | 1 - 2 | 233 | 123 | ||
| GenBank mRNA | AB051517.1 | - | 1 - 2 | 178 | 123 | |
| AK296460.1 | - | 1 - 2 | 233 | 123 | ||
| DQ646393.1 | - | 1 - 2 | 247 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000645.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB051517.1,AK296460.1,DQ646393.1,NM_024646.2,XM_017002336.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010521.hg.1 | TC01000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010521.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010521.hg.1 | ENST00000443756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010521.hg.1 | ENST00000545132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010521.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010520.hg.1 | TC01000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010520.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010520.hg.1 | ENST00000443756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010520.hg.1 | ENST00000545132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010520.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010519.hg.1 | TC01000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010519.hg.1 | ENST00000443756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010519.hg.1 | ENST00000545132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010519.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159321.hg.1 | TC0100008332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159323.hg.1 | TC0100008332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008332.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | ENST00000294353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159320.hg.1 | TC0100008332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159319.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159319.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159319.hg.1 | ENST00000545132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159319.hg.1 | NM_024646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159319.hg.1 | TC0100008332.hg.1 | Clariom D TC |

ZYG11B
Genomic Map