Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008891.hg.1 |
ENST00000502926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008891.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008891.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008891.hg.1 |
NR_001297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012277.hg.1 |
ENST00000624928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010958.hg.1 |
TC05000768.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010958.hg.1 |
ENST00000502926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010958.hg.1 |
NR_001297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012277.hg.1 |
ENST00000624928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
ENST00000502926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
NR_001297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
ENST00000502926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
NR_001297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162022.hg.1 |
TC0500008891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008891.hg.1 |
ENST00000502926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008891.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008891.hg.1 |
ENST00000507007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008891.hg.1 |
NR_001297.1 |
Refseq |