Assay Details
| Gene Symbol: C5orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045116.1 | 1 - 2 | 268 | 99 | ||
| GenBank mRNA | AK096941.1 | - | 1 - 2 | 60 | 99 | |
| AK128882.1 | - | 1 - 2 | 57 | 99 | ||
| AK309191.1 | - | 1 - 2 | 60 | 99 | ||
| BM998490.1 | - | 3 - 4 | 99 | |||
| DC350817.1 | - | 1 - 2 | 94 | 99 | ||
| DN991436.1 | - | 1 - 2 | 276 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096941.1 transcripts.
Set Membership: |
|

C5orf56
Genomic Map