Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000789.hg.1 |
TC08002592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000789.hg.1 |
NR_001578.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000525664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000531764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012265.hg.1 |
ENST00000534302.5 |
Ensembl |