Assay Details
| Gene Symbol: EXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104598.1 | 6 - 7 | 727 | 109 | ||
| XM_017014861.1 | XP_016870350.1 | 4 - 5 | 462 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104598.1 | 6 - 7 | 727 | 109 | ||
| XM_017014861.1 | XP_016870350.1 | 4 - 5 | 462 | 109 | ||
Set Membership: |
|