Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000307544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000376451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000376452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000376454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000376462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000396445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000396446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000442879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000450158.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
ENST00000458595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
NM_001098500.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
NM_019590.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002018.hg.1 |
TC10000169.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000307544.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000376451.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000396445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000396446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
NM_001282769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
NM_001282770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000307544.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000376451.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000396445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
ENST00000396446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
NM_001282769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
NM_001282770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148004.hg.1 |
TC1000007024.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000376451.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000396445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000396446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
NM_001282769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
NM_001282770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000376451.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000396445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
ENST00000396446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
NM_001282769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
NM_001282770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148005.hg.1 |
TC1000007024.hg.1 |
Clariom D TC |