Assay Details
Gene Symbol: PPM1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020700.1 | NP_065751.1 | 1 - 2 | 393 | 116 | |
XM_017019676.1 | XP_016875165.1 | 1 - 2 | 643 | 116 | ||
GenBank mRNA | AB084258.1 | - | 1 - 2 | 658 | 116 | |
AK023315.1 | - | 1 - 2 | 658 | 116 | ||
BC157843.1 | - | 1 - 2 | 403 | 9057062 | 116 | |
DA396561.1 | - | 1 - 2 | 393 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001657.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12022301.hg.1 | ENST00000228705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12022301.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022301.hg.1 | TC12001657.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12022300.hg.1 | ENST00000228705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12022300.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022300.hg.1 | TC12001657.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022302.hg.1 | ENST00000228705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12022302.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022302.hg.1 | TC12001657.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185994.hg.1 | TC1200011054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185998.hg.1 | TC1200011054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185999.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185999.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185999.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185999.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185999.hg.1 | TC1200011054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200186000.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200186000.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200186000.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200186000.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200186000.hg.1 | TC1200011054.hg.1 | Clariom D TC |
