Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022294.hg.1 |
ENST00000228705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022294.hg.1 |
ENST00000551214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022294.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022294.hg.1 |
TC12001657.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
ENST00000619248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185985.hg.1 |
TC1200011054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000228705.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000547857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000548414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000551519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
ENST00000619248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011054.hg.1 |
NM_020700.1 |
Refseq |