Assay Details
Gene Symbol: PPM1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020700.1 | NP_065751.1 | 8 - 9 | 1394 | 127 | |
XM_011538578.2 | XP_011536880.1 | 8 - 9 | 1530 | 127 | ||
XM_011538579.1 | XP_011536881.1 | 8 - 9 | 1082 | 127 | ||
GenBank mRNA | AB032983.1 | - | 7 - 8 | 987 | 127 | |
AB084258.1 | - | 8 - 9 | 1659 | 127 | ||
AK023315.1 | - | 9 - 10 | 1456 | 127 | ||
BC157843.1 | - | 8 - 9 | 1404 | 9057062 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001657.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022292.hg.1 | ENST00000228705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12022292.hg.1 | ENST00000551214.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12022292.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12022292.hg.1 | TC12001657.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185982.hg.1 | TC1200011054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | ENST00000228705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | ENST00000551214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | NM_020700.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200185983.hg.1 | TC1200011054.hg.1 | Clariom D TC |
