Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000216568.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000356921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000357887.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000381269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000394330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000528359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000533541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
ENST00000534137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_001130417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_033262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_058240.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_182932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_182936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
NM_183002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014375.hg.1 |
TC14001265.hg.1 |
HTA TC |