Assay Details
| Gene Symbol: MPV17L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094091.1 | - | 1 - 2 | 766 | 92 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000532896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000533807.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000534421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | ENST00000599612.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007366.hg.1 | NM_032683.2 | Refseq |

MPV17L2
Genomic Map