Assay Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033035.4 | NP_149024.1 | 1 - 2 | 363 | 128 | |
NR_045089.1 | 2 - 3 | 1568 | 128 | |||
GenBank mRNA | AY037115.1 | - | 1 - 2 | 363 | 128 | |
BC040592.1 | - | 2 - 3 | 1569 | 5269764 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000518.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05007202.hg.1 | TC05000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007202.hg.1 | ENST00000420978.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007202.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007206.hg.1 | TC05000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007206.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007206.hg.1 | ENST00000420978.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007206.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007206.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007205.hg.1 | TC05000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007205.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007205.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007204.hg.1 | TC05000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007204.hg.1 | ENST00000420978.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007204.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007207.hg.1 | TC05000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007207.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007207.hg.1 | ENST00000420978.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05007207.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05007207.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156813.hg.1 | TC0500008307.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500156810.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156810.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156810.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156810.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156810.hg.1 | TC0500008307.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000379706.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NM_138551.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008307.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156811.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156811.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156811.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156811.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156811.hg.1 | TC0500008307.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | ENST00000420978.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | NR_045089.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156814.hg.1 | TC0500008307.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500156812.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156812.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156812.hg.1 | ENST00000344895.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156812.hg.1 | NM_033035.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156812.hg.1 | TC0500008307.hg.1 | Clariom D TC |
