Assay Details
| Gene Symbol: SPAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317931.1 | NP_001304860.1 | 6 - 7 | 712 | 125 | |
| NM_003116.2 | NP_003107.1 | 6 - 7 | 731 | 125 | ||
| XM_005260520.4 | XP_005260577.1 | 6 - 7 | 377 | 125 | ||
| XM_011529009.2 | XP_011527311.1 | 6 - 7 | 627 | 125 | ||
| GenBank mRNA | AF262992.1 | - | 6 - 7 | 728 | 125 | |
| BC143783.1 | - | 6 - 7 | 633 | 125 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SPAG4
Genomic Map