Assay Details
Gene Symbol: C11orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005271414.3 | XP_005271471.1 | 9 - 10 | 937 | 205 | |
XM_005271415.4 | XP_005271472.1 | 8 - 9 | 881 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
