Assay Details
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003675.3 | NP_003666.1 | 2 - 3 | 291 | 122 | |
XM_017016860.1 | XP_016872349.1 | 2 - 3 | 419 | 122 | ||
GenBank mRNA | AK314814.1 | - | 2 - 3 | 266 | 122 | |
AU142253.1 | - | 2 - 3 | 291 | 122 | ||
BC000794.1 | - | 2 - 3 | 213 | 3451639 | 122 | |
U51990.1 | - | 2 - 3 | 215 | 122 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000102.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10001146.hg.1 | ENST00000378544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001146.hg.1 | ENST00000378572.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001146.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10001146.hg.1 | TC10000102.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000185089.hg.1 | TC1000012429.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
