Assay Details
| Gene Symbol: ITPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002224.3 | NP_002215.2 | 24 - 25 | 3336 | 91 | |
| XM_011514577.2 | XP_011512879.1 | 23 - 24 | 2976 | 91 | ||
| XM_017010832.1 | XP_016866321.1 | 25 - 26 | 3165 | 91 | ||
| GenBank mRNA | AB209739.1 | - | 15 - 16 | 2123 | 91 | |
| BC146646.1 | - | 24 - 25 | 3295 | 91 | ||
| D26351.1 | - | 24 - 25 | 3347 | 91 | ||
| U01062.1 | - | 24 - 25 | 3148 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000427.hg.1
Set Membership: |
|

ITPR3
Genomic Map