Assay Details
| Gene Symbol: ITPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002224.3 | NP_002215.2 | 29 - 30 | 3995 | 64 | |
| XM_011514577.2 | XP_011512879.1 | 28 - 29 | 3635 | 64 | ||
| XM_017010832.1 | XP_016866321.1 | 30 - 31 | 3824 | 64 | ||
| GenBank mRNA | AB209739.1 | - | 20 - 21 | 2782 | 64 | |
| D26351.1 | - | 29 - 30 | 4006 | 64 | ||
| U01062.1 | - | 29 - 30 | 3807 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000427.hg.1
Set Membership: |
|

ITPR3
Genomic Map