Assay Details
| Gene Symbol: SEMA4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271661.1 | NP_001258590.1 | 3 - 4 | 509 | 82 | |
| XM_017003174.1 | XP_016858663.1 | 2 - 3 | 356 | 82 | ||
| GenBank mRNA | AB022317.1 | - | 3 - 4 | 375 | 82 | |
| AK291023.1 | - | 3 - 4 | 457 | 82 | ||
| BC018361.1 | - | 3 - 4 | 448 | 4215248 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SEMA4F
Genomic Map