| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059579.hg.1 |
TC01003908.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059579.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059579.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059580.hg.1 |
TC01003908.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059580.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059580.hg.1 |
ENST00000497852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01059580.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017575.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232903.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232903.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232903.hg.1 |
ENST00000272164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100232903.hg.1 |
NM_003395.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100232903.hg.1 |
TC0100017575.hg.1 |
Clariom D TC |