Assay Details
Gene Symbol: SOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003578.3 | NP_003569.1 | 1 - 2 | 171 | 118 | |
XM_011538849.2 | XP_011537151.1 | 1 - 2 | 1224 | 118 | ||
XM_011538850.2 | XP_011537152.1 | 1 - 2 | 1224 | 118 | ||
XM_011538851.2 | XP_011537153.1 | 1 - 2 | 1224 | 118 | ||
GenBank mRNA | AF059203.1 | - | 1 - 2 | 134 | 118 | |
BC096090.1 | - | 1 - 2 | 131 | 40000865 | 118 | |
BC096091.3 | - | 1 - 2 | 131 | 40000866 | 118 | |
BC096092.3 | - | 1 - 2 | 131 | 40000868 | 118 | |
BC099626.3 | - | 1 - 2 | 131 | 40000867 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000429.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF059203.1,BC096090.1,BC096091.3,BC096092.3,BC099626.3,NM_003578.3,XM_011538849.2,XM_011538850.2,XM_011538851.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol metabolic process
macrophage derived foam cell differentiation
intestinal cholesterol absorption
cholesterol efflux
very-low-density lipoprotein particle assembly
cholesterol esterification
cholesterol homeostasis
fatty-acyl-CoA binding
cholesterol binding
transferase activity, transferring acyl groups
cholesterol O-acyltransferase activity
cholesterol metabolic process
macrophage derived foam cell differentiation
intestinal cholesterol absorption
cholesterol efflux
very-low-density lipoprotein particle assembly
cholesterol esterification
cholesterol homeostasis
fatty-acyl-CoA binding
cholesterol binding
transferase activity, transferring acyl groups
cholesterol O-acyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12005537.hg.1 | ENST00000301466.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12005537.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12005537.hg.1 | TC12000429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12005534.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12005534.hg.1 | TC12000429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12005536.hg.1 | ENST00000301466.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12005536.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12005536.hg.1 | TC12000429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154990.hg.1 | TC1200007694.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154991.hg.1 | TC1200007694.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007694.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000301466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000542365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | ENST00000551896.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154989.hg.1 | TC1200007694.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200154987.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154987.hg.1 | NM_003578.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200154987.hg.1 | TC1200007694.hg.1 | Clariom D TC |
