Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000330722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000548735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000548735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000548735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000548735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000548735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
ENST00000549898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010758.hg.1 |
NM_005554.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
ENST00000252245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010755.hg.1 |
NM_004693.2 |
Refseq |