Assay Details
Gene Symbol: NT5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002526.3 | NP_002517.1 | 6 - 7 | 1770 | 64 | |
GenBank mRNA | AK075083.1 | - | 3 - 4 | 472 | 64 | |
AK314661.1 | - | 6 - 7 | 1284 | 64 | ||
BC065937.1 | - | 6 - 7 | 1239 | 6163101 | 64 | |
DQ186653.1 | - | 6 - 7 | 1233 | 64 | ||
X55740.1 | - | 6 - 7 | 1263 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000779.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide biosynthetic process
purine nucleotide catabolic process
AMP catabolic process
DNA metabolic process
leukocyte cell-cell adhesion
brain development
response to aluminum ion
dephosphorylation
adenosine biosynthetic process
pyrimidine nucleoside catabolic process
positive regulation of lipid biosynthetic process
negative regulation of inflammatory response
nucleotide binding
ferrous iron binding
5'-nucleotidase activity
purine nucleotide biosynthetic process
purine nucleotide catabolic process
AMP catabolic process
DNA metabolic process
leukocyte cell-cell adhesion
brain development
response to aluminum ion
dephosphorylation
adenosine biosynthetic process
pyrimidine nucleoside catabolic process
positive regulation of lipid biosynthetic process
negative regulation of inflammatory response
nucleotide binding
ferrous iron binding
5'-nucleotidase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009325.hg.1 | TC06000779.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06009325.hg.1 | ENST00000257770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009325.hg.1 | ENST00000369647.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009325.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600160383.hg.1 | TC0600008697.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000257770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369646.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000369651.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000416334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | ENST00000437581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_001204813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008697.hg.1 | NM_002526.3 | Refseq |
