Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009328.hg.1 |
TC06000779.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009328.hg.1 |
ENST00000257770.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009328.hg.1 |
ENST00000369651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009328.hg.1 |
NM_001204813.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009328.hg.1 |
NM_002526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000257770.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000369651.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000416334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
NM_001204813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
NM_002526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000257770.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000369651.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
ENST00000416334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
NM_001204813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
NM_002526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160385.hg.1 |
TC0600008697.hg.1 |
Clariom D TC |