Assay Details
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015.4 | NP_001006.1 | 1 - 2 | 107 | 60 | |
| GenBank mRNA | AK311809.1 | - | 1 - 2 | 42 | 60 | |
| AV759412.2 | - | 1 - 2 | 48 | 60 | ||
| BC007283.1 | - | 1 - 2 | 66 | 3343839 | 60 | |
| BC007603.1 | - | 1 - 2 | 66 | 3350204 | 60 | |
| BC007945.2 | - | 1 - 2 | 67 | 4297932 | 60 | |
| BC010028.2 | - | 1 - 2 | 66 | 3357773 | 60 | |
| BC018829.2 | - | 1 - 2 | 66 | 60 | ||
| BC100025.1 | - | 1 - 2 | 117 | 30565084 | 60 | |
| BI550467.1 | - | 1 - 2 | 43 | 60 | ||
| X06617.1 | - | 1 - 2 | 30 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000735.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK311809.1,BC007283.1,BC007603.1,BC007945.2,BC010028.2,BC100025.1,NM_001015.4,X06617.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
osteoblast differentiation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
osteoblast differentiation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19010293.hg.1 | ENST00000270625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010293.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010293.hg.1 | TC19000735.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010297.hg.1 | ENST00000270625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010297.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010297.hg.1 | TC19000735.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000363660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008542.hg.1 | NR_001285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009507.hg.1 | ENST00000488441.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017929.hg.1 | ENST00000420757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006993.hg.1 | ENST00000442125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006993.hg.1 | ENST00000442125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009507.hg.1 | ENST00000488441.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000363660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008542.hg.1 | NR_001285.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | TC1900008542.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000594493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000596873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000599561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000601216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000601306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | ENST00000602252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166106.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017929.hg.1 | ENST00000420757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | NM_001015.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | TC1900008542.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000270625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000599167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | ENST00000600027.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166095.hg.1 | NM_001015.4 | Refseq |

RPS11
Genomic Map