| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008967.hg.1 |
ENST00000305904.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008967.hg.1 |
ENST00000338542.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008967.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008967.hg.1 |
TC16000633.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163876.hg.1 |
TC1600008559.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600163865.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163865.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163865.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600163865.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600163865.hg.1 |
TC1600008559.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000305904.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000338542.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000561880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000562982.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000568035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
ENST00000570222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_001305017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NM_130897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008559.hg.1 |
NR_130943.1 |
Refseq |