Assay Details
| Gene Symbol: HEATR5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019024.2 | NP_061897.1 | 18 - 19 | 2798 | 133 | |
| XM_006712034.2 | XP_006712097.1 | 18 - 19 | 2737 | 133 | ||
| XM_006712035.3 | XP_006712098.1 | 18 - 19 | 2801 | 133 | ||
| XM_011532934.2 | XP_011531236.1 | 18 - 19 | 3265 | 133 | ||
| XM_011532935.2 | XP_011531237.1 | 7 - 8 | 937 | 133 | ||
| XM_017004378.1 | XP_016859867.1 | 18 - 19 | 2737 | 133 | ||
| XM_017004379.1 | XP_016859868.1 | 19 - 20 | 3175 | 133 | ||
| XM_017004380.1 | XP_016859869.1 | 19 - 20 | 2948 | 133 | ||
| GenBank mRNA | AB037835.1 | - | 9 - 10 | 1245 | 133 | |
| AK096637.1 | - | 18 - 19 | 2737 | 133 | ||
| BC150605.1 | - | 18 - 19 | 2783 | 9056975 | 133 | |
| BX538008.1 | - | 12 - 13 | 1721 | 133 | ||
| BX648805.1 | - | 4 - 5 | 958 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001738.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027950.hg.1 | TC02001738.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027950.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027950.hg.1 | ENST00000354531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027950.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189388.hg.1 | TC0200012256.hg.1 | Clariom D TC |

HEATR5B
Genomic Map