Assay Details
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022128.2 | NP_071411.1 | 1 - 2 | 139 | 117 | |
GenBank mRNA | AJ404857.1 | - | 1 - 2 | 99 | 117 | |
AK300989.1 | - | 1 - 2 | 93 | 117 | ||
AY643715.1 | - | 1 - 2 | 131 | 117 | ||
BC017425.1 | - | 1 - 2 | 141 | 4694872 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001695.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02027411.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027411.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027411.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027411.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027415.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027415.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027415.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027415.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027412.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027412.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027412.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027412.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188406.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188412.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188410.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 162809 | - | - |
