Assay Details
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287580.1 | NP_001274509.1 | 3 - 4 | 1389 | 133 | |
NM_022128.2 | NP_071411.1 | 2 - 3 | 270 | 133 | ||
GenBank mRNA | AJ404857.1 | - | 2 - 3 | 230 | 133 | |
AK300989.1 | - | 2 - 3 | 224 | 133 | ||
BC017425.1 | - | 2 - 3 | 272 | 4694872 | 133 | |
BX648909.1 | - | 3 - 4 | 1360 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001695.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001287580.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02027410.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027410.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027410.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027410.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027409.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027409.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027409.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027409.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188408.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188408.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188408.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188408.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188408.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188404.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188405.hg.1 | TC0200012098.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000536835.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006691.hg.1 | ENST00000560994.1 | Ensembl |
