Assay Details
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287580.1 | NP_001274509.1 | 6 - 7 | 1680 | 106 | |
NM_022128.2 | NP_071411.1 | 5 - 6 | 561 | 106 | ||
GenBank mRNA | AJ404857.1 | - | 5 - 6 | 521 | 106 | |
AK300989.1 | - | 5 - 6 | 515 | 106 | ||
AY643715.1 | - | 5 - 6 | 553 | 106 | ||
BC017425.1 | - | 5 - 6 | 563 | 4694872 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001695.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027406.hg.1 | TC02001695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027406.hg.1 | ENST00000302188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027406.hg.1 | ENST00000444339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027406.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000453412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | ENST00000612015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012098.hg.1 | NR_028308.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000302188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000449378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | ENST00000458185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | NM_001287580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | NM_022128.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188400.hg.1 | TC0200012098.hg.1 | Clariom D TC |
