| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8071714 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8071713 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
214623_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11760550_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11760551_x_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11760551_PM_x_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11760550_at |
- |
- |
| Affymetrix Human Clariom D Assay |
PSR2200147122.hg.1 |
TC2200006836.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200147122.hg.1 |
ENST00000426721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147122.hg.1 |
NR_033408.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200147122.hg.1 |
ENST00000426721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147122.hg.1 |
NR_033408.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200147121.hg.1 |
TC2200006836.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200147121.hg.1 |
ENST00000426721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147121.hg.1 |
NR_033408.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200147121.hg.1 |
ENST00000426721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147121.hg.1 |
NR_033408.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
214623_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214623_at |
- |
- |
| Human Genome U133A 2.0 Array |
214623_at |
- |
- |
| Human Genome U133A 2.0 Array |
214623_PM_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw513707 |
- |
- |
| Human Genome U95 Set |
517_PM_at |
- |
- |
| Human Genome U95 Set |
517_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214623_at9 |
- |
- |
| Human Genome U95 Set |
76675_PM_f_at |
- |
- |
| Human Genome U95 Set |
76675_f_at |
- |
- |
| Human Genome U133 Set |
214623_at |
- |
- |
| Human Genome U133 Set |
214623_PM_at |
- |
- |