Assay Details
| Gene Symbol: MRPL40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003776.3 | NP_003767.2 | 1 - 2 | 702 | 70 | |
| GenBank mRNA | AF034091.1 | - | 1 - 2 | 61 | 70 | |
| AJ295637.1 | - | 1 - 2 | 59 | 70 | ||
| AK123768.1 | - | 1 - 2 | 702 | 70 | ||
| BC009707.1 | - | 1 - 2 | 91 | 3876531 | 70 | |
| CR456532.1 | - | 1 - 2 | 54 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000044.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22000362.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000362.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000362.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000362.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000360.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000360.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000360.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000360.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000357.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000357.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000357.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000363.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000363.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000363.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000356.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000356.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000356.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000361.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000361.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000361.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000361.hg.1 | TC22000044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145164.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145159.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145159.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145159.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145159.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145159.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006601.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145165.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145168.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145161.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145166.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | TC2200006601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | NM_003776.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000333130.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000443660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | ENST00000471259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145167.hg.1 | NM_003776.3 | Refseq |

MRPL40
Genomic Map