Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
TC01002293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
ENST00000375217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
ENST00000375224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
ENST00000375226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
ENST00000375254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
ENST00000375267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035014.hg.1 |
NM_020765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
ENST00000375224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
ENST00000375254.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
NM_020765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
ENST00000375224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
ENST00000375254.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
NM_020765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196995.hg.1 |
TC0100013123.hg.1 |
Clariom D TC |