Assay Details
| Gene Symbol: TPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000366.5 | NP_000357.3 | 4 - 5 | 686 | 154 | |
| NM_001018006.1 | NP_001018006.1 | 4 - 5 | 686 | 154 | ||
| NM_001018020.1 | NP_001018020.1 | 4 - 5 | 686 | 154 | ||
| XM_005254639.3 | XP_005254696.2 | 4 - 5 | 1129 | 154 | ||
| XM_005254646.2 | XP_005254703.1 | 3 - 4 | 512 | 154 | ||
| XM_005254650.3 | XP_005254707.1 | 3 - 4 | 512 | 154 | ||
| XM_005254651.1 | XP_005254708.1 | 3 - 4 | 512 | 154 | ||
| XM_005254652.1 | XP_005254709.1 | 3 - 4 | 512 | 154 | ||
| XM_005254653.2 | XP_005254710.1 | 3 - 4 | 512 | 154 | ||
| XM_006720667.3 | XP_006720730.2 | 4 - 5 | 1129 | 154 | ||
| XM_017022536.1 | XP_016878025.1 | 4 - 5 | 1129 | 154 | ||
| XM_017022537.1 | XP_016878026.1 | 4 - 5 | 641 | 154 | ||
| XM_017022538.1 | XP_016878027.1 | 4 - 5 | 1129 | 154 | ||
| XM_017022539.1 | XP_016878028.1 | 4 - 5 | 1129 | 154 | ||
| GenBank mRNA | AF474156.1 | - | 3 - 4 | 288 | 154 | |
| AK092051.1 | - | 3 - 4 | 1403 | 154 | ||
| AK092573.1 | - | 2 - 3 | 1508 | 154 | ||
| AK298625.1 | - | 3 - 4 | 402 | 154 | ||
| AK309170.1 | - | 4 - 5 | 895 | 154 | ||
| BC007433.2 | - | 4 - 5 | 636 | 2962925 | 154 | |
| BX458841.2 | - | 4 - 5 | 555 | 154 | ||
| DQ424903.1 | - | 3 - 4 | 387 | 154 | ||
| GQ472198.1 | - | 4 - 5 | 636 | 154 | ||
| KU178366.1 | - | 4 - 5 | 495 | 154 | ||
| M19715.1 | - | 3 - 4 | 332 | 154 | ||
| Z24727.1 | - | 710 | 154 | |||
More Information
Set Membership: |
|

TPM1
Genomic Map