Assay Details
| Gene Symbol: TPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018004.1 | NP_001018004.1 | 8 - 9 | 966 | 139 | |
| NM_001018006.1 | NP_001018006.1 | 8 - 9 | 966 | 139 | ||
| NM_001018007.1 | NP_001018007.1 | 8 - 9 | 966 | 139 | ||
| NM_001018020.1 | NP_001018020.1 | 8 - 9 | 966 | 139 | ||
| NM_001301289.1 | NP_001288218.1 | 7 - 8 | 806 | 139 | ||
| XM_005254637.1 | XP_005254694.1 | 9 - 10 | 1047 | 139 | ||
| XM_005254652.1 | XP_005254709.1 | 7 - 8 | 792 | 139 | ||
| GenBank mRNA | AJ000147.1 | - | 1 - 2 | 67 | 139 | |
| AJ001055.1 | - | 1 - 2 | 67 | 139 | ||
| AK057989.1 | - | 10 - 11 | 752 | 139 | ||
| AK092051.1 | - | 6 - 7 | 2624 | 139 | ||
| AK092573.1 | - | 7 - 8 | 1864 | 139 | ||
| AK131384.1 | - | 9 - 10 | 980 | 139 | ||
| AK301319.1 | - | 7 - 8 | 702 | 139 | ||
| AL050179.1 | - | 8 - 9 | 847 | 139 | ||
| M19715.1 | - | 7 - 8 | 612 | 139 | ||
More Information
Set Membership: |
|

TPM1
Genomic Map