Assay Details
| Gene Symbol: RPS27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002954.5 | NP_002945.1 | 1 - 2 | 304 | 142 | |
| GenBank mRNA | BM907473.1 | - | 5 - 6 | 142 | ||
| CR746519.1 | - | 1 - 2 | 315 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_002954.5 transcripts.
Set Membership: |
|

RPS27A
Genomic Map