Assay Details
| Gene Symbol: HEATR5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019024.2 | NP_061897.1 | 33 - 34 | 5644 | 140 | |
| XM_006712035.3 | XP_006712098.1 | 33 - 34 | 5647 | 140 | ||
| XM_011532934.2 | XP_011531236.1 | 33 - 34 | 6111 | 140 | ||
| XM_011532935.2 | XP_011531237.1 | 22 - 23 | 3783 | 140 | ||
| XM_017004378.1 | XP_016859867.1 | 32 - 33 | 5316 | 140 | ||
| XM_017004379.1 | XP_016859868.1 | 34 - 35 | 6021 | 140 | ||
| XM_017004380.1 | XP_016859869.1 | 34 - 35 | 5794 | 140 | ||
| GenBank mRNA | AB037835.1 | - | 24 - 25 | 4091 | 140 | |
| AK001513.1 | - | 6 - 7 | 1084 | 140 | ||
| AK026928.1 | - | 10 - 11 | 1904 | 140 | ||
| AL832880.1 | - | 6 - 7 | 1046 | 140 | ||
| BC150605.1 | - | 33 - 34 | 5629 | 9056975 | 140 | |
| BX538008.1 | - | 27 - 28 | 4567 | 140 | ||
| BX648805.1 | - | 19 - 20 | 3804 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001738.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027931.hg.1 | TC02001738.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027931.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027931.hg.1 | ENST00000354531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027931.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000425467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000467978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000471051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | ENST00000478810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012256.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189365.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189365.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189365.hg.1 | ENST00000233099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189365.hg.1 | NM_019024.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189365.hg.1 | TC0200012256.hg.1 | Clariom D TC |

HEATR5B
Genomic Map