Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
ENST00000325084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
ENST00000357018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
ENST00000360228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
ENST00000418012.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
NM_000068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
NM_001127221.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
NM_001127222.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
NM_001174080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
NM_023035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017149.hg.1 |
TC19001220.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000360228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000573710.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000585802.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000586190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000587525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000614285.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_000068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001127221.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001127222.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001174080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_023035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
TC1900011894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000360228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000573710.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000585802.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000586190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000587525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
ENST00000614285.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_000068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001127221.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001127222.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_001174080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204849.hg.1 |
NM_023035.2 |
Refseq |