Assay Details
| Gene Symbol: GLDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000170.2 | NP_000161.2 | 9 - 10 | 1456 | 96 | |
| GenBank mRNA | AK310534.1 | - | 5 - 6 | 831 | 96 | |
| AK314156.1 | - | 9 - 10 | 1458 | 96 | ||
| BC111993.1 | - | 9 - 10 | 1329 | 8327461 | 96 | |
| BC111995.1 | - | 9 - 10 | 1329 | 8327463 | 96 | |
| D90239.1 | - | 9 - 10 | 1413 | 96 | ||
| M63635.1 | - | 9 - 10 | 1344 | 96 | ||
| M64590.1 | - | 9 - 10 | 1413 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GLDC
Genomic Map