Assay Details
Gene Symbol: CITED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168388.2 | NP_001161860.1 | 1 - 2 | 92 | 73 | |
GenBank mRNA | BG900380.1 | - | 1 - 2 | 129 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002168.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001168388.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
liver development
outflow tract morphogenesis
regulation of organ formation
transcription, DNA-templated
determination of left/right symmetry
heart development
sex determination
cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell-cell adhesion
cell differentiation
negative regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
response to fluid shear stress
positive regulation of peroxisome proliferator activated receptor signaling pathway
adrenal cortex formation
negative regulation of apoptotic process
response to estrogen
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
spleen development
ventricular septum morphogenesis
embryonic heart tube left/right pattern formation
left/right pattern formation
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
left/right axis specification
nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
positive regulation of male gonad development
RNA polymerase II transcription coactivator activity
RNA polymerase II transcription corepressor activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
histone acetyltransferase binding
LBD domain binding
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
liver development
outflow tract morphogenesis
regulation of organ formation
transcription, DNA-templated
determination of left/right symmetry
heart development
sex determination
cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell-cell adhesion
cell differentiation
negative regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
response to fluid shear stress
positive regulation of peroxisome proliferator activated receptor signaling pathway
adrenal cortex formation
negative regulation of apoptotic process
response to estrogen
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
spleen development
ventricular septum morphogenesis
embryonic heart tube left/right pattern formation
left/right pattern formation
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
left/right axis specification
nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
positive regulation of male gonad development
RNA polymerase II transcription coactivator activity
RNA polymerase II transcription corepressor activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
histone acetyltransferase binding
LBD domain binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | TC06002168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | ENST00000367651.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | ENST00000392312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | ENST00000536159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | ENST00000537332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027310.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | TC06002168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | ENST00000367651.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | ENST00000392312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | ENST00000536159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | ENST00000537332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027312.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | TC06002168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | ENST00000367651.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | ENST00000536159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | ENST00000537332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027311.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027315.hg.1 | TC06002168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027315.hg.1 | ENST00000536159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027315.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192422.hg.1 | TC0600013361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600192427.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192427.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192427.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192427.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192427.hg.1 | TC0600013361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192421.hg.1 | TC0600013361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013361.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000367651.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000536159.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000537332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | ENST00000618718.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_001168388.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_001168389.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | NM_006079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192423.hg.1 | TC0600013361.hg.1 | Clariom D TC |
