Assay Details
| Gene Symbol: PRPF40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031698.2 | NP_001026868.2 | 11 - 12 | 1117 | 118 | |
| NM_012272.2 | NP_036404.1 | 10 - 11 | 1407 | 118 | ||
| XM_006719324.3 | XP_006719387.1 | 10 - 11 | 2171 | 118 | ||
| XM_006719325.3 | XP_006719388.1 | 10 - 11 | 2171 | 118 | ||
| XM_011538135.2 | XP_011536437.1 | 10 - 11 | 2171 | 118 | ||
| XM_011538136.2 | XP_011536438.1 | 10 - 11 | 2171 | 118 | ||
| XM_011538137.2 | XP_011536439.1 | 9 - 10 | 2090 | 118 | ||
| XM_011538138.2 | XP_011536440.1 | 9 - 10 | 2060 | 118 | ||
| XM_011538139.2 | XP_011536441.1 | 11 - 12 | 1111 | 118 | ||
| XM_011538140.2 | XP_011536442.1 | 11 - 12 | 1111 | 118 | ||
| XM_011538141.1 | XP_011536443.1 | 10 - 11 | 937 | 118 | ||
| XM_011538143.2 | XP_011536445.1 | 10 - 11 | 953 | 118 | ||
| XM_011538144.2 | XP_011536446.1 | 9 - 10 | 826 | 118 | ||
| XM_017019135.1 | XP_016874624.1 | 10 - 11 | 874 | 118 | ||
| XM_017019136.1 | XP_016874625.1 | 9 - 10 | 826 | 118 | ||
| XM_017019137.1 | XP_016874626.1 | 9 - 10 | 826 | 118 | ||
| XM_017019138.1 | XP_016874627.1 | 10 - 11 | 2171 | 118 | ||
| XM_017019139.1 | XP_016874628.1 | 6 - 7 | 837 | 118 | ||
| GenBank mRNA | AL834216.1 | - | 10 - 11 | 1407 | 118 | |
| BC031917.1 | - | 8 - 9 | 713 | 118 | ||
| BC050398.2 | - | 11 - 12 | 953 | 118 | ||
| BC067364.1 | - | 10 - 11 | 938 | 6644672 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019139.1 transcripts.
Set Membership: |
|

PRPF40B
Genomic Map