Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004684.hg.1 |
ENST00000261897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004684.hg.1 |
ENST00000380281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004684.hg.1 |
NM_001031698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004684.hg.1 |
NM_012272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004684.hg.1 |
TC12000377.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004685.hg.1 |
ENST00000261897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004685.hg.1 |
ENST00000380281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004685.hg.1 |
NM_001031698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004685.hg.1 |
NM_012272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004685.hg.1 |
TC12000377.hg.1 |
HTA TC |