Assay Details
| Gene Symbol: CEACAM22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027754.2 | 2 - 3 | 504 | 137 | ||
| GenBank mRNA | AK123850.1 | - | 2 - 3 | 504 | 137 | |
| AK302997.1 | - | 3 - 4 | 430 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001609.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302997.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19022011.hg.1 | ENST00000446628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022011.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022011.hg.1 | TC19001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022012.hg.1 | ENST00000446628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022012.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022012.hg.1 | TC19001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022010.hg.1 | ENST00000446628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022010.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022010.hg.1 | TC19001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190553.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190553.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190553.hg.1 | TC1900010897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190553.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190553.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | TC1900010897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190551.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010897.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | TC1900010897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190543.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | TC1900010897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000455058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190546.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | NR_027754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | TC1900010897.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | ENST00000446628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | ENST00000455455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190552.hg.1 | NR_027754.2 | Refseq |

CEACAM22P
Genomic Map