Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
TC01002827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
ENST00000294661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
ENST00000471115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044001.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044003.hg.1 |
TC01002827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044003.hg.1 |
ENST00000294661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044003.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044003.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210068.hg.1 |
TC0100014774.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210066.hg.1 |
TC0100014774.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000294661.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000344356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000471115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
ENST00000493514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NM_198077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014774.hg.1 |
NR_024113.1 |
Refseq |