| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
TC0X000495.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006577.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
TC0X000495.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006578.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008001.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154576.hg.1 |
TC0X00008001.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
ENST00000372656.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
ENST00000372661.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006612.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_001006614.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
NM_016303.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154577.hg.1 |
TC0X00008001.hg.1 |
Clariom D TC |