Assay Details
| Gene Symbol: C2orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099334.2 | NP_001092804.1 | 3 - 4 | 294 | 120 | |
| XM_017004074.1 | XP_016859563.1 | 3 - 4 | 307 | 120 | ||
| XM_017004075.1 | XP_016859564.1 | 3 - 4 | 242 | 120 | ||
| XM_017004076.1 | XP_016859565.1 | 3 - 4 | 282 | 120 | ||
| XM_017004077.1 | XP_016859566.1 | 3 - 4 | 242 | 120 | ||
| XM_017004078.1 | XP_016859567.1 | 3 - 4 | 242 | 120 | ||
| GenBank mRNA | BC035737.1 | - | 3 - 4 | 294 | 5730487 | 120 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002725.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043746.hg.1 | TC02002725.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043746.hg.1 | ENST00000341287.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043746.hg.1 | ENST00000451346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043746.hg.1 | ENST00000453017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043746.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043743.hg.1 | TC02002725.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043743.hg.1 | ENST00000341287.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043743.hg.1 | ENST00000451346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043743.hg.1 | ENST00000453017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043743.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214283.hg.1 | TC0200015575.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214280.hg.1 | TC0200015575.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000341287.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000423952.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000428015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000449053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000451346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | ENST00000453017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015575.hg.1 | NM_001099334.2 | Refseq |

C2orf80
Genomic Map