Assay Details
Gene Symbol: C11orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135635.1 | NP_001129107.1 | 1 - 2 | 150 | 52 | |
GenBank mRNA | CX866110.1 | - | 1 - 2 | 126 | 52 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001949.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11023403.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023403.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023403.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023403.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023403.hg.1 | TC11001949.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023400.hg.1 | TC11001949.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011260.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186608.hg.1 | TC1100011260.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186607.hg.1 | TC1100011260.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000438576.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000449692.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | ENST00000530188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | NM_001135635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | NM_031450.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100186604.hg.1 | TC1100011260.hg.1 | Clariom D TC |
