Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014402.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014402.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014402.hg.1 |
TC10001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014403.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014403.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014403.hg.1 |
TC10001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014404.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014404.hg.1 |
TC10001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014400.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014400.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014400.hg.1 |
TC10001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000464386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000464386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169425.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169425.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169425.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169425.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169425.hg.1 |
TC1000010192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169426.hg.1 |
TC1000010192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169428.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169428.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169428.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169428.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169428.hg.1 |
TC1000010192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169427.hg.1 |
TC1000010192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000375377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000464386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000464386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
ENST00000465712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010192.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169429.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169429.hg.1 |
NM_020848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169429.hg.1 |
TC1000010192.hg.1 |
Clariom D TC |