Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000227.hg.1 |
ENST00000412293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000227.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000227.hg.1 |
TC16000012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000435229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000226.hg.1 |
ENST00000412293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000226.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000226.hg.1 |
TC16000012.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144325.hg.1 |
TC1600006459.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144324.hg.1 |
TC1600006459.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000435229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006459.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
ENST00000412293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
ENST00000457760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
NR_024453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144326.hg.1 |
TC1600006459.hg.1 |
Clariom D TC |