Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291333.1 | NP_001278262.1 | 1 - 2 | 194 | 121 | |
| NM_004623.4 | NP_004614.3 | 1 - 2 | 194 | 121 | ||
| GenBank mRNA | AF063602.1 | - | 1 - 2 | 156 | 121 | |
| AF073887.1 | - | 1 - 2 | 140 | 121 | ||
| AK025524.1 | - | 1 - 2 | 194 | 121 | ||
| AK222562.1 | - | 1 - 2 | 147 | 121 | ||
| AK292426.1 | - | 1 - 2 | 155 | 121 | ||
| BC001276.1 | - | 1 - 2 | 133 | 3462738 | 121 | |
| BT006817.1 | - | 1 - 2 | 107 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TTC4
Genomic Map