Assay Details
| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036625.1 | 1 - 2 | 317 | 128 | ||
| GenBank mRNA | DA072915.1 | - | 1 - 2 | 130 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VDAC1| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036625.1 | 1 - 2 | 317 | 128 | ||
| GenBank mRNA | DA072915.1 | - | 1 - 2 | 130 | 128 | |
Set Membership: |
|